Description
- Two-part, white, thermally conductive silicone
- Cures with the addition of heat
- 20:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide heat transfer between electrical/electronic components and their heat sinks
- Use to adhere integrated circuit substrates, base plates, heat sinks or where grooves or other configurations require a non-flowable to limited flow material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.64 W/(mK) |
Cure System | Platinum |
Tensile | 425 psi (2.93 MPa) |
Appearance | White |
Cure | 4 hours / 65 °C |
Durometer | 60 Type A |
Elongation | 250 % |
Lap Shear | 275 psi (1.9 MPa) |
Mix Ratio | 20:1 |
Specific Gravity | 1.27 |
Tear | 50 ppi (8.82 kN/m) |
Work Time | 2 hours |
Comment | Primed w/CF1-135, Contains Boron Nitride |