Description
- Two-part, white, thermally conductive silicone
- Cures with the addition of heat
- Mix Ratio 10:1 (Part A:B)
- Designed for enhanced performance in extreme low and high temperatures
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide moderate heat transfer between electrical/electronic components and their heat sinks
- Use for adhering openings in modules and housings where grooves or other configurations require a limited flow material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 0.79 W/(mK) |
Viscosity | 300,000 cP (300,000 mPa*s) |
Cure System | Platinum |
Tensile | 275 psi (1.9 MPa) |
Appearance | White |
Cte | |
Cure | 30 minutes / 150 °C |
Durometer | 75 Type A |
Elongation | 40 % |
Lap Shear | 205 psi (1.41 MPa) |
Mix Ratio | 10:1 |
Specific Gravity | 1.38 |
Tack Free Time | 3.5 hours |
Tear | 45 ppi (7.94 kN/m) |
Work Time | 2 hours |
Comment | *22) 0.8 W/mK Low Viscosity, *3) Primed Lap Shear 205 psi (1.4 MPa), Low Temp |