Description
- Two-part, white, thermally conductive silicone
- Cures with the addition of heat
- 15:1 Mix Ratio (Part A:B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide heat transfer between electrical/electronic components and their heat sinks
- Use to adhere integrated circuit substrates, base plates, heat sinks or where grooves or other configurations require a non-flowable to limited flow material
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 1.49 W/(mK) |
Viscosity | 3,560,000 cP (3,560,000 mPa*s) |
Cure System | Platinum |
Tensile | 200 psi (1.38 MPa) |
Appearance | White |
Cure | 7 days / RTV |
Dielectric Strength | 540 volts/mil (21.06 kV/mm) |
Durometer | 75 Type A |
Elongation | 30 % |
Lap Shear | 160 psi (1.1 MPa) |
Mix Ratio | 15:1 |
Specific Gravity | 1.53 |
Tack Free Time | 4.5 hours |
Tear | 50 ppi (8.82 kN/m) |
Volume Resistivity | 530,000,000,000,000 ohmcm |
Work Time | 2 hours |
Comment | *22) 1.5 W/mK *,4 ) Primed Lap Shear 160 psi (1.1 MPa) |