Description
- Two-part, gray, thermally conductive RTV silicone
- 100:0.2 Mix Ratio (Base: Curing Agent)
- Designed for enhanced performance in extreme low and high temperatures
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide moderate heat transfer between electrical/electronic components and their heat sinks
- Use to adhere openings in modules or housings where grooves and other configurations require a limited flow material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 1.22 W/(mK) |
Cure System | Alkoxy |
Tensile | 750 psi (5.17 MPa) |
Appearance | Gray |
Cte | |
Cure | 7 days / RTV |
Durometer | 90 Type A |
Elongation | 35 % |
Lap Shear | 475 psi (3.28 MPa) |
Mix Ratio | 100:0.2 |
Specific Gravity | 2.55 |
Tear | 90 ppi (15.87 kN/m) |
Work Time | 2 hours |
Comment | *22) 1.2 W/mK, *3) Primed Lap Shear 475 psi (3.3 MPa) |