Description
- Two-part, gray, thermally conductive silicone
- Based on a dimethyl silicone polymer
- Uses a platinum-catalyzed addition cure
- 20:1 Mix Ratio (Part A:B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- To provide moderate heat transfer between electrical/electronic components and their heat sinks
- Use to adhere openings in modules or housings where grooves and other configurations require a non-flowable to limited flow material
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 1 W/(mK) |
Cure System | Platinum |
Tensile | 650 psi (4.48 MPa) |
Appearance | Gray |
Cte | |
Cure | 24 hours / RTV |
Dielectric Strength | 430 volts/mil (16.77 kV/mm) |
Durometer | 85 Type A |
Elongation | 15 % |
Lap Shear | 375 psi (2.59 MPa) |
Mix Ratio | 20:1 |
Specific Gravity | 2.4 |
Tack Free Time | 4 hours |
Tear | 55 ppi (9.7 kN/m) |
Volume Resistivity | 140,000,000,000,000 ohmcm |
Work Time | 2.5 hours |
Comment | *22) 1.0 W/mK, *4) Primed Lap Shear 375 psi (2.6 MPa) |