Description
- Two-part film adhesive
- Cures at room temperature or rapidly with heat
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- For bonding or sealing silicone elastomers and some metals or plastics
- For applications requiring shorter work times, easy clean-up, and consistent bond thickness
Product Details
Property | Average Result |
---|---|
Appearance | Translucent |
Cte | |
Cure | 4 hours / 65 °C |
Cure System | Platinum |
Cure Type | RTV |
Elongation | 850 % |
Lap Shear | 250 psi (1.7 MPa) |
Rheology | Thixotropic / Non-slump |
Substrate | Primerless: ABS / Aluminum / Glass / Metal / PEEK / Plastic / PMMA / Polycarbonate / Polyetherimide / Polymide / Polyurethane / Silicone / Stainless steel / Titanium |
Tear | 115 ppi (20.3 kN/m) |
Tensile | 1,450 psi (10.0 MPa) |