Description
- Two-part, electrically conductive film adhesive
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- Use for RFI and EMI shielding in electrical and space applications
- For bonding or sealing silicone elastomers and some metals or plastics
- For applications requiring shorter work times, easy clean-up, and consistent bond thickness
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Appearance | Black |
Cure | 4 hours / 65 °C |
Volume Resistivity | 150 ohmcm |
Comment | Use for RFI and EMI shielding in electrical and space applications. For applications requiring shorter work times, easy clean-up, and consistent bond thickness. |