Description
- Two-part film adhesive
- Cures at room temperature or rapidly with heat
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- For bonding or sealing silicone elastomers and some metals or plastics
- For applications requiring shorter work times, easy clean-up, and consistent bond thickness.
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Lap Shear | 250 psi (1.72 MPa) |
Appearance | Translucent |
Application Life | 7 minutes |
Cte | |
Cure | 4 hours / 65 °C |
Elongation | 400 % |
Tear | 115 ppi (20.28 kN/m) |
Tensile | 1,450 psi (10 MPa) |
Comment | Low Outgassing version of R-2682-12. |