Description
- Two-part, tan-colored electrically conductive RTV silicone
- 100:0.5 Mix Ratio (Base: Curing Agent)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use for RFI and EMI shielding in electrical and space applications
- Use to adhere covers onto housings or for any application where grooves and other configurations require a non-flowable to limited flow material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Volume Resistivity | 0.01 ohmcm |
Cure System | Alkoxy |
Durometer | 80 Type A |
Tensile | 400 psi (2.76 MPa) |
Appearance | Tan |
Cte | |
Cure | 10 days / RTV |
Elongation | 90 % |
Lap Shear | 325 psi (2.24 MPa) |
Mix Ratio | 100:0.5 |
Specific Gravity | 3.86 |
Tear | 60 ppi (10.58 kN/m) |
Thermal Conductivity | 1 W/(mK) |
Work Time | 3.5 hours |
Comment | Electrically Conductive, 0.007 ohmcm |