Description
- Two-part, tan-colored electrically conductive silicone
- 20:1 Mix Ratio (Part A:B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use for RFI and EMI shielding in electrical and space applications
- Well suited for form-in-place conductive gaskets
Product Details
Property | Average Result |
---|---|
Volume Resistivity | 0.01 ohmcm |
Viscosity | 224,500 cP (224,500 mPa*s) |
Cure System | Platinum |
Durometer | 85 Type A |
Tensile | 525 psi (3.62 MPa) |
Appearance | Tan |
Cte | |
Cure | 30 minutes / 150 °C |
Elongation | 86 % |
Mix Ratio | 20:1 |
Specific Gravity | 3.39 |
Thermal Conductivity | 1.2 W/(mK) |
Work Time | 2.75 hours |
Comment | 0.005 ohmcm, *22) 1.2 W/mK |