Description
- A low density, two-part, red, thixotropic RTV silicone
- 100: 0.5 Mix Ratio (Base: Curing Agent)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- As a bonding, sealing, or potting material in electronic and space applications
- Provides radiation resistance, low thermal conductivity, oxidation stability, thermal stability and good ablative characteristics
- Especially useful to bond solar cells to solar array panels
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Alkoxy |
Viscosity | 125,000 cP (125,000 mPa*s) |
Lap Shear | 100 psi (0.69 MPa) |
Durometer | 50 Type A |
Appearance | Red |
Cte | |
Cure | 7 days / RTV |
Dielectric Strength | 645 volts/mil (25.16 kV/mm) |
Elongation | 60 % |
Mix Ratio | 100:0.5 |
Specific Gravity | 0.64 |
Tack Free Time | 13 hours |
Tensile | 175 psi (1.21 MPa) |
Work Time | 4 hours |
Comment | Long Worktime, Low Density* 1, )Primed Lap Shear 100 psi (0.69 MPa) |