Description
- Two-part, flowable, red RTV silicone
- 100: 0.5 Mix Ratio (Base: Curing Agent)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- As a sealing, caulking, adhesive or potting material in electronic and space applications where high vacuum environments are encountered
- For bonding solar cells to solar array panels
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Alkoxy |
Viscosity | 55,000 cP (55,000 mPa*s) |
Lap Shear | 500 psi (3.45 MPa) |
Durometer | 55 Type A |
Appearance | Rust Red |
Cte | |
Cure | 7 days / RTV |
Dielectric Strength | 825 volts/mil (32.18 kV/mm) |
Elongation | 150 % |
Mix Ratio | 100:0.5 |
Specific Gravity | 1.49 |
Tack Free Time | 8 hours |
Tear | 40 ppi (7.05 kN/m) |
Tensile | 950 psi (6.55 MPa) |
Thermal Conductivity | 0.27 W/(mK) |
Work Time | 3 hours |
Comment | Pourable RTV Cure, * 1 )Primed Lap Shear 500 psi (3.4 MPa) |