Description
- Two-part, optically clear silicone system
- 10:1 mix ratio (Part A:B)
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- As an embedding or potting compound for environmental protection of electronic assemblies and components
- Provides protection from humidity, radiation, thermal stress and mechanical stress
- As an adhesive in applications such as solar cell arrays where clarity and low volatility are important
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 8,000 cP (8,000 mPa*s) |
Lap Shear | 400 psi (2.76 MPa) |
Durometer | 50 Type A |
Appearance | Clear |
Cure | 15 minutes / 150 °C |
Elongation | 125 % |
Mix Ratio | 10:1 |
Specific Gravity | 1.02 |
Tack Free Time | 10 hours |
Tensile | 1,000 psi (6.9 MPa) |
Work Time | 2 hours |
Comment | Pourable, Optically Clear |