Description
- Two-part, low viscosity, black silicone system
- Offers a broad operating temperature range
- 10:1 mix ratio (Part A:B)
Applications
- For electronic and space applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
- As an embedding or potting compound for environmental protection of electronic assemblies and components
- Provides protection from extremes in temperature, humidity, radiation, thermal stress and mechanical stress
- As an adhesive in optical applications requiring a black material to block light
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 8,400 cP (8,400 mPa*s) |
Durometer | 50 Type A |
Appearance | Black |
Cte | |
Cure | 30 minutes / 150 °C |
Dielectric Strength | 850 volts/mil (33.15 kV/mm) |
Elongation | 150 % |
Mix Ratio | 10:1 |
Specific Gravity | 1.05 |
Tack Free Time | 6 hours |
Tensile | 950 psi (6.55 MPa) |
Work Time | 3 hours |
Comment | Low Viscosity, Fast Cure |