Description
- Yields a low to medium density silicone rubber foam when rapidly catalyzed
- Low viscosity
- 1:10 Mix Ratio (Part A: Part B)
Applications
- In shock and vibration dampening situations requiring a lightweight, flexible foam
- For intricate back-filling due to a low viscosity
- For applications requiring low outgassing and minimal volatile condensable materials.
Product Details
Property | Average Result |
---|---|
Viscosity | 3,000 cP (3,000 mPa*s) |
Cure System | Platinum |
Foam Density | 20 lbs/ft3 (0.32 g/cm3) |
Application Life | 6 minutes |
Appearance | White |
Cure | 60 minutes / RTV |
Mix Ratio | 1:10 |
Comment | Cure in place versatile low outgassing foam, application life 6 minutes |