Description
- Two-part, translucent silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 50,000 cP (50,000 mPa*s) |
Lap Shear | 400 psi (2.76 MPa) |
Durometer | 30 Type A |
Appearance | Translucent |
Cure | 15 minutes / 150 °C |
Elongation | 350 % |
Mix Ratio | 1:1 |
Tensile | 750 psi (5.17 MPa) |
Work Time | 30 minutes |
Comment | Tough, self leveling and good adhesion to various metals. Recommended for broad temperature applications. 400 psi (2.8 MPa), primed (CF1-135) lap shear. |