CV-2289-2
CONTROLLED VOLATILITY POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
- Two-part, black silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For use as an adhesive
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 52,500 cP (52,500 mPa*s) |
Lap Shear | 425 psi (2.93 MPa) |
Durometer | 30 Type A |
Appearance | Black |
Cure | 15 minutes / 150 °C |
Elongation | 400 % |
Mix Ratio | 1:1 |
Tack Free Time | 5 hours |
Tear | 50 ppi (8.82 kN/m) |
Tensile | 750 psi (5.17 MPa) |
Work Time | 30 minutes |
Comment | Pourable Elastomer |