Description
- Two-part, white silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For use as an adhesive
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Rheology | Flowable / Self Leveling |
Work Time | 30 minutes |
Durometer | 34 Type A |
Appearance | White |
Cure | 15 minutes / 150 °C |
Cure Type | Heat Cure / RTV |
Dielectric Strength | 955 volts/mil (37.25 kV/mm) |
Elongation | 343 % |
Lap Shear | 375 psi (2.6 MPa) |
Mix Ratio | 1:1 |
Rheology | Flowable / Self Leveling |
Specific Gravity | 1.12 |
Tack Free Time | 5 hours |
Tensile | 720 psi (5.0 MPa) |
Viscosity | 50,000 cP (50,000 mPa*s) |