Description
- Two-part, translucent, pourable silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- 10:1 Mix Ratio (Part A:Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 85,000 cP (85,000 mPa*s) |
Lap Shear | 450 psi (3.1 MPa) |
Durometer | 30 Type A |
Appearance | Translucent |
Cte | |
Cure | 30 minutes / 150 °C |
Dielectric Strength | 900 volts/mil (35.1 kV/mm) |
Elongation | 400 % |
Mix Ratio | 10:1 |
Specific Gravity | 1.11 |
Tear | 60 ppi (10.58 kN/m) |
Tensile | 750 psi (5.17 MPa) |
Work Time | 3.5 hours |
Comment | Low / High Temperature, Flowable, Fast Cure |