Description
- Two-part, translucent, pourable silicone system
- Offers a high tear strength, good physical properties
- 10:1 Mix Ratio (Part A:B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to a high tear strength
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 90,000 cP (90,000 mPa*s) |
Durometer | 35 Type A |
Appearance | Translucent |
Cure | 15 minutes / 150 °C |
Elongation | 400 % |
Mix Ratio | 10:1 |
Specific Gravity | 1.1 |
Tack Free Time | 15 hours |
Tear | 75 ppi (13.23 kN/m) |
Tensile | 925 psi (6.38 MPa) |
Work Time | 3 hours |
Comment | Tough, self leveling and good adhesion to various metals. |