Description
- One-part, flat black silicone dispersion
- Provided as an RTV silicone dispersion in VM&P Naphtha
Applications
- For space and electrical applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- Provides atomic oxygen protection and resistance to radiation and thermal stress
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Viscosity | 2,400 cP (2,400 mPa*s) |
Non Volatile Content | 72 % |
Appearance | Black |
Cure | 7 days / RTV |
Cure System | Oxime |
Dielectric Strength | 845 volts/mil (32.96 kV/mm) |
Mix Ratio | 1 PART |
Specific Gravity | 1.26 |
Comment | 72% Solids |