Description
- One-Part, translucent silicone RTV
- Non-slumping in thin sections
Applications
- For applications requiring extreme low temperature, low outgassing and minimal volatile condensables under extreme operating conditions
- As a sealing, caulking, adhesive or potting material in electronics and space applications requiring minimal outgassing to avoid condensation in sensitive devices
- For bonding and sealing in applications such as overhead or vertical joints that require a non-slumping and one-part material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Cure System | Oxime |
Extrusion Rate | 35 g/minute |
Lap Shear | 375 psi (2.59 MPa) |
Durometer | 45 Type A |
Appearance | Translucent |
Cte | |
Cure | 7 days / RTV |
Dielectric Strength | 1,100 volts/mil (42.9 kV/mm) |
Elongation | 300 % |
Mix Ratio | 1 PART |
Refractive Index | 1.43 |
Specific Gravity | 1.11 |
Tack Free Time | 20 minutes |
Tensile | 700 psi (4.83 MPa) |
Comment | For applications requiring a broader operating temperature range. Available in black and white. 200 psi (1.4 MPa), Unprimed lap shear. |