CareSil
CSF-3504
GUM BLEND
Description
  • Film Forming
  • Semi-occlusive
  • High Purity; <0.05% D4 cyclic content
  • Delivers a rich feel and a powdery after-feel
  • Helps in pigment dispersion
  • Increased substantivity without Cyclopentasiloxane
  • Increased shine
Applications
  • Moisturization
  • Anti-Aging/Anti-Wrinkle
  • Sunscreen, After-Sun, and Self-Tanners
  • Skin Repair Serums
  • Skin Tone Repair
  • Men’s Grooming
  • Color Cosmetics
Product Details
Property Average Result
INCI Name Dimethicone (and) Dimethiconol
D4 Content <0.05 %
Viscosity 1,140,000 cP (1,140,000 mPa*s)
Appearance Clear
Refractive Index 1.40
Specific Gravity 0.94
Comment 50% High Purity Dimethiconol Dispersed in Low Viscosity Dimethicone Fluid