CF6-135

HIGH TECHNOLOGY SILICONE PRIMER

Description

  • Specially formulated primer designed for use with platinum-cured systems where conventional silicone primers are insufficient
  • Translucent to preserve visibility with translucent products
  • One-component primers supplied in VM&P Naphtha require no mixing
  • Air-drying
  • Convenient container sizes produce less waste

Applications

  • Improves the adhesion of addition-cured systems to various substrates including: metals, glass, ceramics, some plastics and other silicone material

Product Details

Property Average Result
Non Volatile Content 9 %
Specific Gravity 0.78
Appearance Translucent
Cure System Condensation
Comment Recommended where platinum inhibition is of concern.

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn more

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.