CareSil
CES-1104
ENCAPSULATION TECHNOLOGY
Description
  • Elastomer gel delivered from an hydrophilic gel
  • Surfactant free, silicone membrane stabilization
  • Cyclic free
  • Ease of use and dispersion: directly in the water phase
  • Light and Smooth feel for aqueous base formulations
  • Transformable texture: light and watery at the beginning that turns into a matte velvet veil
  • With carefully controlled conditions, possible to formulate transparent products
Applications
  • Moisturization
  • Anti-Aging/Anti-Wrinkle
  • Sunscreen, After-Sun, and Self-Tanners
  • Skin Repair Serums
  • Skin Tone Repair
  • Men's Grooming
  • Color Cosmetics
Product Details
Property Average Result
Inci Name
D4 Content <0.05 %
Viscosity 60,000 cP (60,000 mPa*s)
Appearance Translucent
Refractive Index 1.40
Specific Gravity 1.03
Comment Encapsulated Silicone with 12% CXG-1104