Premium Care Line
MED-4901
LIQUID SILICONE RUBBER
Description
  • Two-part, translucent silicone system designed for use with injection molding equipment
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For the injection molding of parts requiring a material with a very low durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings
  • and other precision parts
  • Suitable for over-molding applications
  • Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
NuSil Technology's MED-4901 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 40 Type 00
Tensile 285 psi (1.97 MPa)
Elongation 1135 %
Tear 55 ppi (9.7 kN/m)
Appearance Translucent
Cure 5 minutes / 150 °C
Cure Rate Scorch 0.9 minutes
Cure Rate T90 1.2 minutes
Cure System Platinum
Extrusion Rate 160 g/minute
Mix Ratio 1:1
Specific Gravity 1.1
Stress at Strain 15 psi / 300 % (0.1 Mpa / 300 %)
Work Time 25 hours
Comment Injection Molding Elastomer