Description
- A very soft two-part, low viscosity, silicone elastomer
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 55 Type 000 |
Viscosity | 6,450 cP (6,450 mPa*s) |
Work Time | 18 hours |
Tensile | 40 psi (0.28 MPa) |
Cure | 45 minutes / 150 °C |
Cure System | Platinum |
Elongation | 475 % |
Mix Ratio | 1:1 |
Stress at Strain | 8 psi / 200 % (0.06 MPa / 200 %) |
Comment | Ultra Soft Elastomer |